Learn More
Medchemexpress LLC Atovaquone-d4 | 2070015-14-2 | 99.8% | 370.86 | 1 MG
SDP
Catalog No. 5000350114
Encompass_Preferred
Atovaquone-d4 is the deuterium labeled Atovaquone, a medication used to treat or prevent pneumocystis pneumonia, toxoplasmosis, malaria, and babesia. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-50114 Supplier Medchemexpress LLC Supplier No. HY13832S1MG
Atovaquone-d4 is the deuterium labeled Atovaquone, a medication used to treat or prevent pneumocystis pneumonia, toxoplasmosis, malaria, and babesia. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS