Learn More
Medchemexpress LLC ATP-13C2 dilithium (Adenosine 5'-triphosphate-13C2 dilithium) | 56-65-5 | 96.0% | 1 MG
SDP
Catalog No. 5000412340
Encompass_Preferred
ATP-13C2 (Adenosine 5'-triphosphate-13C2) dilithium is a 13C-labeled ATP (HY-B2176). ATP (Adenosine 5'-triphosphate) is a key component of energy storage and metabolism in living organisms. It supplies metabolic energy for metabolic pumps and functions as a coenzyme in cells. ATP is also a significant endogenous signaling molecule in immunity and inflammation.
  • This compound can be used as a tracer.
  • It can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-12340 Supplier Medchemexpress LLC Supplier No. HYB2176S71MG
ATP-13C2 (Adenosine 5'-triphosphate-13C2) dilithium is a 13C-labeled ATP (HY-B2176). ATP (Adenosine 5'-triphosphate) is a key component of energy storage and metabolism in living organisms. It supplies metabolic energy for metabolic pumps and functions as a coenzyme in cells. ATP is also a significant endogenous signaling molecule in immunity and inflammation.
  • This compound can be used as a tracer.
  • It can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.