Learn More
TARGETMOL CHEMICALS INC ATP 1G
SDP
Catalog No. 502877772
Encompass_Preferred
Also available in 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. ATP (Adenosine triphosphate) provides cellular energy, participates in overall energy balance, and maintains intracellular homeostasis. ATP can act as an extracellular signaling molecule through interactions with specific purinergic receptors to mediate a variety of processes including neurotransmission, inflammation, apoptosis, and bone remodeling. Purity 98.87%
Catalog No. 50-287-7772 Supplier TARGETMOL CHEMICALS INC Supplier No. T200891G
Also available in 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. ATP (Adenosine triphosphate) provides cellular energy, participates in overall energy balance, and maintains intracellular homeostasis. ATP can act as an extracellular signaling molecule through interactions with specific purinergic receptors to mediate a variety of processes including neurotransmission, inflammation, apoptosis, and bone remodeling. Purity 98.87%