Learn More
Selleck Chemical LLC ATP S5260-1g
SDP
Catalog No. 503112761
Encompass_Preferred
ATP (Adenosine-Triphosphate Adenosine 5 -triphosphate) is a multifunctional nucleoside triphosphate and an important endogenous signaling molecule in immunity and inflammation
Catalog No. 50-311-2761 Supplier Selleck Chemical LLC Supplier No. S52601G
ATP (Adenosine-Triphosphate Adenosine 5 -triphosphate) is a multifunctional nucleoside triphosphate and an important endogenous signaling molecule in immunity and inflammation