Learn More
Medchemexpress LLC HY-B2176 500mg , ATP CAS:56-65-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB2176500MG
Encompass_Preferred
HY-B2176 500mg ATP CAS: 56-65-5 ATP (Adenosine 5'-triphosphate) is a central component of energy storage and metabolism in vivo. ATP provides the metabolic energy to drive metabolic pumps and serves as a coenzyme in cells. ATP is an important endogenous signaling molecule in immunity and inflammation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9820