Learn More
Bioworld ATP, Disodium Trihydrate, 100 g
Catalog No. 50255373
Encompass_Preferred
Specifications:Em (259nm, phosphate buffer, pH 7.0): >14700Ultra pure gradeA250/A260: 0.75-0.81A280/A260: 0.114-0.18Heavy Metals (as Pb): <0.002%ADP and ADP: <0.5%Appearance: Solid.ChEBI : Not available.Crystal_Structure : Not available.InChI : Not available.InChIKey : Not available.Melting Point: 188-190 °CRTECS Number: AU7417000
Catalog No. 50-255-373 Supplier Bioworld Supplier No. 401900202
Specifications:Em (259nm, phosphate buffer, pH 7.0): >14700Ultra pure gradeA250/A260: 0.75-0.81A280/A260: 0.114-0.18Heavy Metals (as Pb): <0.002%ADP and ADP: <0.5%Appearance: Solid.ChEBI : Not available.Crystal_Structure : Not available.InChI : Not available.InChIKey : Not available.Melting Point: 188-190 °CRTECS Number: AU7417000