Learn More
Dojindo Molecular Technologies Inc ATP Assay Kit-Luminescence (50 tests) is used to determine intracellular levels of ATP by luciferase luminescence assay. High-sensitivity detection can be performed with stable luminescence. 50 tests
SDP
Catalog No. NC1933303
Encompass_Preferred
Adenosine triphosphate (ATP) is an energy source for living cells and is synthesized by both glycolysis and mitochondrial oxidative phosphorylation. Decreased ATP levels are associated with cancer, aging, neurodegenerative diseases, and mitochondrial diseases. The ATP Assay Kit-Luminescence enables the quantitation of intracellular ATP by luciferase luminescence assay. This kit can be applied for microplate assay. Without a complicated process including medium removal and cell washing, the ATP Assay Kit-Luminescence can generate a stable luminescent signal with a half-life of greater than 3 hours by simply mixing the kit components and adding the reagent. This kit contains an ATP Standard, which provides highly reproducible data without measuring unstable ATP. Dojindo has various quantification kits of Intracellular Metabolites. (https://www.dojindo.com/quantification-of-intracellular-metabolites/)
Catalog No. NC1933303 Supplier Dojindo Molecular Technologies Inc Supplier No. A55010
Adenosine triphosphate (ATP) is an energy source for living cells and is synthesized by both glycolysis and mitochondrial oxidative phosphorylation. Decreased ATP levels are associated with cancer, aging, neurodegenerative diseases, and mitochondrial diseases. The ATP Assay Kit-Luminescence enables the quantitation of intracellular ATP by luciferase luminescence assay. This kit can be applied for microplate assay. Without a complicated process including medium removal and cell washing, the ATP Assay Kit-Luminescence can generate a stable luminescent signal with a half-life of greater than 3 hours by simply mixing the kit components and adding the reagent. This kit contains an ATP Standard, which provides highly reproducible data without measuring unstable ATP. Dojindo has various quantification kits of Intracellular Metabolites. (https://www.dojindo.com/quantification-of-intracellular-metabolites/)