Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC ATP Citrate Lyase Antibody (YA596) | 121 kDa | 10 UL
SDP

Catalog No. 5000396944
Encompass_Preferred

ATP Citrate Lyase Antibody (YA596) is a rabbit host antibody reactive with human and mouse samples. It is supplied as a liquid at 1 mg/mL and is suitable for various applications including Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunoprecipitation, and Flow Cytometry.

  • Rabbit host antibody
  • Reacts with human and mouse samples
  • Supplied as a liquid at 1 mg/mL
  • Applications: Western Blot, ICC/IF, IHC-P, IP, FC
  • Formulated in 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol
  • Contains 0.05% Sodium Azide preservative
  • Stored at -20°C for 1 year
  • Avoid repeated freeze / thaw cycles

Catalog No. 50-003-96944 Supplier Medchemexpress LLC Supplier No. HYP8038010UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ATP Citrate Lyase Antibody (YA596) is a rabbit host antibody reactive with human and mouse samples. It is supplied as a liquid at 1 mg/mL and is suitable for various applications including Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunoprecipitation, and Flow Cytometry.

  • Rabbit host antibody
  • Reacts with human and mouse samples
  • Supplied as a liquid at 1 mg/mL
  • Applications: Western Blot, ICC/IF, IHC-P, IP, FC
  • Formulated in 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol
  • Contains 0.05% Sodium Azide preservative
  • Stored at -20°C for 1 year
  • Avoid repeated freeze / thaw cycles

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.