Learn More
TRIBIOSCIENCE INC ATP Colorimetric/Fluorometric Assay Kit, 100 Tests
SDP
Supplier:  TRIBIOSCIENCE INC TBS2010
Encompass_Preferred
ATP Colorimetric and Fluorometric Assay kit can detect down to 0.5 uM of ATP in various samples.
Catalog No. NC2886224