Learn More
Medchemexpress LLC HY-B0345A 1g , ATP (disodium salt) CAS:987-65-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0345A1G
Encompass_Preferred
HY-B0345A 1g ATP (disodium salt) CAS: 987-65-5 ATP disodium salt (Adenosine 5'-triphosphate disodium salt) is a central component of energy storage and metabolism in vivo, provides the metabolic energy to drive metabolic pumps and serves as a coenzyme in cells. ATP disodium salt is an important endogenous signaling molecule in immunity and inflammation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1130