Learn More
Apexbio Technology LLC ATP disodium salt 987-65-5 10mM (in 1mL H2O)
SDP
Catalog No. 5000064247
Encompass_Preferred
ATP disodium salt (CAS 987-65-5) is a nucleotide derivative serving as the primary energy carrier in cellular metabolism It provides chemical energy through phosphate group transfer reactions facilitating numerous biochemical processes In addition to its metabolic functions ATP acts as a coenzyme involved in intracellular signaling and protein phosphorylation pathways Due to these biochemical roles ATP disodium salt is widely utilized in research exploring cellular metabolism signaling mechanisms and energy regulation pathways Recently ATP s function as a signaling molecule in immune responses and inflammation modulation has also gained attention
Catalog No. 50-000-64247 Supplier Apexbio Technology LLC Supplier No. B33045.1
ATP disodium salt (CAS 987-65-5) is a nucleotide derivative serving as the primary energy carrier in cellular metabolism It provides chemical energy through phosphate group transfer reactions facilitating numerous biochemical processes In addition to its metabolic functions ATP acts as a coenzyme involved in intracellular signaling and protein phosphorylation pathways Due to these biochemical roles ATP disodium salt is widely utilized in research exploring cellular metabolism signaling mechanisms and energy regulation pathways Recently ATP s function as a signaling molecule in immune responses and inflammation modulation has also gained attention