Learn More
Selleck Chemical LLC ATP disodium trihydrate E4964-100MG
SDP
Catalog No. 5000041163
Encompass_Preferred
ATP disodium trihydrate is the primary energy currency of the cell essential for intracellular energy metabolism It regulates extracellular biological processes including cardiac function neurotransmission muscle contraction vasodilation bone and liver glycogen metabolism and inflammation It also plays a crucial role in initiating immune responses during infections
Catalog No. 50-000-41163 Supplier Selleck Chemical LLC Supplier No. E4964100MG
ATP disodium trihydrate is the primary energy currency of the cell essential for intracellular energy metabolism It regulates extracellular biological processes including cardiac function neurotransmission muscle contraction vasodilation bone and liver glycogen metabolism and inflammation It also plays a crucial role in initiating immune responses during infections