Learn More
Gold Biotechnology Inc ATP Disodium trihydrate
SDP
Catalog No. 5000562077
Encompass_Preferred
Adenosine-5 -triphosphate (ATP) is an important molecule in vivo for energy storage and metabolism and is often called the molecular unit of currency of intracellular energy transfer ATP is a coenzyme in multiple reactions as well as a substrate for multiple kinases where it helps to propagate a signal either inside or outside of the cell
Catalog No. 50-005-62077 Supplier Gold Biotechnology Inc Supplier No. A0811
Adenosine-5 -triphosphate (ATP) is an important molecule in vivo for energy storage and metabolism and is often called the molecular unit of currency of intracellular energy transfer ATP is a coenzyme in multiple reactions as well as a substrate for multiple kinases where it helps to propagate a signal either inside or outside of the cell