Learn More
Enzo Life Sciences Adenosine-5'-O-(3-thiotriphosphate). tetralithium salt (25 mg). CAS: 93839-89-5
SDP
Supplier:  Enzo Life Sciences BMLST1010025
Encompass_Preferred
An ATP analog that can substitute for ATP in various kinase reactions. The resulting thiophosphorylated proteins are resistant to dephosphorylation by protein phosphatases. Alternative name: ATPγ-S . 4Li. Purity: ≥85% (HPLC). Solubility: Soluble in water (25mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-2149