Learn More
Abcam ATP-gamma-S, Kinase substrate, 100UG
Catalog No. 502583463
Encompass_Preferred
ATP-gamma-S is a kinase substrate and inhibitor of ATP-dependent enzyme systems. Hydrolyzed very slowly by phosphatases and most ATPases. Once thiophosphorylated, proteins are resistant to protein phosphatases. P2 purinergic receptor agonist. - Cited in over 30 publications - Purity > 80% - Soluble in water to 10 mM
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-3463 Supplier Abcam Supplier No. AB138911100UG
ATP-gamma-S is a kinase substrate and inhibitor of ATP-dependent enzyme systems. Hydrolyzed very slowly by phosphatases and most ATPases. Once thiophosphorylated, proteins are resistant to protein phosphatases. P2 purinergic receptor agonist. - Cited in over 30 publications - Purity > 80% - Soluble in water to 10 mM
The product is subject to the following: Abcam Restricted Use Statement