Learn More
Biotium ATP-Glo™ Bioluminometric Cell Viability Assay Kit (1000 assays)
Catalog No. 501964476
Encompass_Preferred
ATP-Glo™ Bioluminometric Cell Viability Assay offers a highly sensitive assay for quantifying ATP. The homogeneous assay procedure involves a single addition of ATP-Glo™ Detection Cocktail directly to cells cultured in a serum-supplemented medium. It is not necessary to remove cell culture medium or wash cells before adding the reagent. This ATP detection kit takes advantage of firefly luciferase’s use of ATP to oxidize D-Luciferin and the resulting production of light in order to assess the amount of ATP available. The ATP-Glo™ kit can be used to detect as little as a single cell or 0.01 picomole of ATP. The signal produced is linear within 6 orders of magnitude. By relating the amount of ATP to the number of viable cells, the assay has wide applications, ranging from the determination of viable cell numbers to cell proliferation to cell cytotoxicity. This kit has reagents sufficient for 1000 assays in a 96-well format.
Catalog No. 50-196-4476 Supplier Biotium Supplier No. 300202
ATP-Glo™ Bioluminometric Cell Viability Assay offers a highly sensitive assay for quantifying ATP. The homogeneous assay procedure involves a single addition of ATP-Glo™ Detection Cocktail directly to cells cultured in a serum-supplemented medium. It is not necessary to remove cell culture medium or wash cells before adding the reagent. This ATP detection kit takes advantage of firefly luciferase’s use of ATP to oxidize D-Luciferin and the resulting production of light in order to assess the amount of ATP available. The ATP-Glo™ kit can be used to detect as little as a single cell or 0.01 picomole of ATP. The signal produced is linear within 6 orders of magnitude. By relating the amount of ATP to the number of viable cells, the assay has wide applications, ranging from the determination of viable cell numbers to cell proliferation to cell cytotoxicity. This kit has reagents sufficient for 1000 assays in a 96-well format.