Learn More
Medchemexpress LLC ATP-polyamine-biotin | 1800401-93-7 | 98.1% | 5 MG
SDP
Catalog No. 5000410753
Encompass_Preferred
ATP-polyamine-biotin is the first cell-permeable ATP analogue, functioning as an efficient kinase cosubstrate. This compound is capable of promoting the biotin labeling of kinase substrates within live cells.
  • First cell-permeable ATP analogue
  • Efficient kinase cosubstrate
  • Promotes biotin labeling of kinase substrates in live cells
Catalog No. 50-004-10753 Supplier Medchemexpress LLC Supplier No. HYD01835MG
ATP-polyamine-biotin is the first cell-permeable ATP analogue, functioning as an efficient kinase cosubstrate. This compound is capable of promoting the biotin labeling of kinase substrates within live cells.
  • First cell-permeable ATP analogue
  • Efficient kinase cosubstrate
  • Promotes biotin labeling of kinase substrates in live cells