Learn More
Medchemexpress LLC ATP-polyamine-biotin | 1800401-93-7 | MFCD29924712 | >98.0% | 945.85 g/mol | C32H58N11O14P3S | 10 MG
SDP
Catalog No. 5000297344
Encompass_Preferred
ATP-polyamine-biotin is a cell-permeable ATP analogue used as an efficient kinase co-substrate to transfer a biotin tag to kinase substrates in vitro and in live cells. It facilitates detection and enrichment of phosphorylated proteins for downstream analysis in biochemical and cell-based assays.
  • Promotes biotin labeling of kinase substrates in live cells.
  • Functions as an efficient ATP cosubstrate for kinases.
  • Enables detection and enrichment of phosphorylated proteins.
  • Provided with high purity suitable for biochemical assays.
  • Available in multiple pack sizes, including 10 mg.
Catalog No. 50-002-97344 Supplier Medchemexpress LLC Supplier No. HYD018310MG
ATP-polyamine-biotin is a cell-permeable ATP analogue used as an efficient kinase co-substrate to transfer a biotin tag to kinase substrates in vitro and in live cells. It facilitates detection and enrichment of phosphorylated proteins for downstream analysis in biochemical and cell-based assays.
  • Promotes biotin labeling of kinase substrates in live cells.
  • Functions as an efficient ATP cosubstrate for kinases.
  • Enables detection and enrichment of phosphorylated proteins.
  • Provided with high purity suitable for biochemical assays.
  • Available in multiple pack sizes, including 10 mg.