Learn More
Medchemexpress LLC ATP-Red 1 | 1847485-97-5 | 561.48 | 10 MG
SDP
Catalog No. 5000447580
Encompass_Preferred
ATP-Red 1 is a multisite-binding switchable fluorescent probe designed to selectively and rapidly respond to intracellular concentrations of ATP in living cells, with an excitation/emission wavelength of 510/590 nm. It is for research use only and not intended for human use.
  • Selectively and rapidly responds to intracellular concentrations of ATP in living cells
  • Excitation/emission wavelength of 510/590 nm
  • Used for measuring ATP production in HeLa cells
  • Used for staining mitochondrial ATP in neurons
  • Used for assessing ATP content in H9C2 and HK-2 cells
  • Used for detecting ATP content in SNU-368 cells
Catalog No. 50-004-47580 Supplier Medchemexpress LLC Supplier No. HYU0045110MG
ATP-Red 1 is a multisite-binding switchable fluorescent probe designed to selectively and rapidly respond to intracellular concentrations of ATP in living cells, with an excitation/emission wavelength of 510/590 nm. It is for research use only and not intended for human use.
  • Selectively and rapidly responds to intracellular concentrations of ATP in living cells
  • Excitation/emission wavelength of 510/590 nm
  • Used for measuring ATP production in HeLa cells
  • Used for staining mitochondrial ATP in neurons
  • Used for assessing ATP content in H9C2 and HK-2 cells
  • Used for detecting ATP content in SNU-368 cells