Learn More
Medchemexpress LLC ATP-Red 1 | 1847485-97-5 | 561.48 | 25 MG
SDP
Catalog No. 5000447739
Encompass_Preferred
ATP-Red 1 is a multisite-binding switchable fluorescent probe that selectively and rapidly responds to intracellular concentrations of ATP in living cells.
  • Multisite-binding switchable fluorescent probe
  • Selectively and rapidly responds to intracellular concentrations of ATP in living cells
  • Excitation/Emission wavelengths (Ex/Em) = 510/590 nm
Catalog No. 50-004-47739 Supplier Medchemexpress LLC Supplier No. 25MGHYU00451
ATP-Red 1 is a multisite-binding switchable fluorescent probe that selectively and rapidly responds to intracellular concentrations of ATP in living cells.
  • Multisite-binding switchable fluorescent probe
  • Selectively and rapidly responds to intracellular concentrations of ATP in living cells
  • Excitation/Emission wavelengths (Ex/Em) = 510/590 nm