Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC ATP-Red 1 | 1847485-97-5 | 561.48 | 280.74 UG
SDP

Catalog No. 5000447684
Encompass_Preferred

ATP-Red 1 is a multisite-binding switchable fluorescent probe that selectively and rapidly responds to intracellular concentrations of ATP in living cells (Ex/Em = 510/590 nm). It is for research use only.

  • Selectively and rapidly responds to intracellular ATP in living cells.
  • Multisite-binding switchable fluorescent probe.
  • Excitation/Emission at 510/590 nm.
  • Solid appearance, pale purple to purple color.
  • Molecular formula: C34H36BN3O4.
  • Recommended storage: 4°C, protect from light.
  • In solvent storage: -80°C for 6 months; -20°C for 1 month (protect from light).
  • Solubility in DMSO: 50 mg/mL (89.05 mM).

Catalog No. 50-004-47684 Supplier Medchemexpress LLC Supplier No. HYU00451Y280.74UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ATP-Red 1 is a multisite-binding switchable fluorescent probe that selectively and rapidly responds to intracellular concentrations of ATP in living cells (Ex/Em = 510/590 nm). It is for research use only.

  • Selectively and rapidly responds to intracellular ATP in living cells.
  • Multisite-binding switchable fluorescent probe.
  • Excitation/Emission at 510/590 nm.
  • Solid appearance, pale purple to purple color.
  • Molecular formula: C34H36BN3O4.
  • Recommended storage: 4°C, protect from light.
  • In solvent storage: -80°C for 6 months; -20°C for 1 month (protect from light).
  • Solubility in DMSO: 50 mg/mL (89.05 mM).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.