Learn More
Medchemexpress LLC HY-U00451 5mg Medchemexpress, ATP-Red 1 CAS:1847485-97-5 Purity:>98%
SDP
Catalog No. 501934458
Encompass_Preferred
Medchemexpress, HY-U00451 5mg ATP-Red 1 CAS:1847485-97-5 ATP-Red 1 is a multisite-binding switchable fluorescent probe, and can selectively and rapidly responds to intracellular concentrations of ATP in living cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4458 Supplier Medchemexpress LLC Supplier No. HYU004515MG
Medchemexpress, HY-U00451 5mg ATP-Red 1 CAS:1847485-97-5 ATP-Red 1 is a multisite-binding switchable fluorescent probe, and can selectively and rapidly responds to intracellular concentrations of ATP in living cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.