Learn More
Medchemexpress LLC ATP-Red 1 (solution) | 1847485-97-5 | 561.48 | 56.14 UG
SDP
Catalog No. 5000446919
Encompass_Preferred
ATP-Red 1 is a multisite-binding switchable fluorescent probe designed for the selective and rapid detection of intracellular ATP concentrations in living cells. It features excitation at 510 nm and emission at 590 nm, making it suitable for various cell-staining applications.
  • Selectively responds to intracellular ATP
  • Rapid detection in living cells
  • Excitation at 510 nm
  • Emission at 590 nm
  • Suitable for fluorescence microscopy or flow cytometry
Catalog No. 50-004-46919 Supplier Medchemexpress LLC Supplier No. HYU00451Y56.14UG
ATP-Red 1 is a multisite-binding switchable fluorescent probe designed for the selective and rapid detection of intracellular ATP concentrations in living cells. It features excitation at 510 nm and emission at 590 nm, making it suitable for various cell-staining applications.
  • Selectively responds to intracellular ATP
  • Rapid detection in living cells
  • Excitation at 510 nm
  • Emission at 590 nm
  • Suitable for fluorescence microscopy or flow cytometry