Learn More
Enzo Life Sciences ATP (Energy) Regeneration Solution (100µl)
SDP
Catalog No. 502010788
Encompass_Preferred
This solution contains MgCl2, ATP, and ATP regenerating enzymes to recycle ATP hydrolysis products, (AMP, ADP) to ATP. An ideal energy source for conjugation/degradation assays (ubiquitin, proteasome, SUMO, etc.) particularly those that are preformed with crude lysates/impure reagents that may contain ATP-depleting activities. Formulation: Liquid. In 50mM TRIS-Cl, pH 7.5. Long Term Storage: -80°C.
Catalog No. 50-201-0788 Supplier Enzo Life Sciences Supplier No. BMLEW98100100
Add to Cart
Add to Cart
This solution contains MgCl2, ATP, and ATP regenerating enzymes to recycle ATP hydrolysis products, (AMP, ADP) to ATP. An ideal energy source for conjugation/degradation assays (ubiquitin, proteasome, SUMO, etc.) particularly those that are preformed with crude lysates/impure reagents that may contain ATP-depleting activities. Formulation: Liquid. In 50mM TRIS-Cl, pH 7.5. Long Term Storage: -80°C.