Learn More
Apexbio Technology LLC ATP Solution (100 mM) 1mL
SDP
Catalog No. 503107275
Encompass_Preferred
ATP (Adenosine-5 -triphosphate) known as in Chinese is a fundamental component for energy transfer in biological systems ATP and its sodium salts can be utilized in numerous molecular biology applications such as kinase reactions in vitro transcription ligation reactions and phosphorylation assays This product is a colorless transparent aqueous solution prepared from ATP trisodium salt with a purity 99% (by HPLC) concentration of 100 mM pH (25 ) 7 0 0 1 and free of DNase RNase and phosphatase contamination
Catalog No. 50-310-7275 Supplier Apexbio Technology LLC Supplier No. K10431
ATP (Adenosine-5 -triphosphate) known as in Chinese is a fundamental component for energy transfer in biological systems ATP and its sodium salts can be utilized in numerous molecular biology applications such as kinase reactions in vitro transcription ligation reactions and phosphorylation assays This product is a colorless transparent aqueous solution prepared from ATP trisodium salt with a purity 99% (by HPLC) concentration of 100 mM pH (25 ) 7 0 0 1 and free of DNase RNase and phosphatase contamination