Learn More
Lonza Walkersville ATP Standard, 5 mL
Catalog No. NC0015934
Encompass
The ATP Standard is a specialized aqueous preparation of adenosine triphosphate (ATP) and is primarily intended for use in research to calibrate ATP assays based on the luciferase bioluminescence technique. Each vial contains 5 ml of 10 µM ATP.Content:1 x Bottle ATP Standard, 5 mL
Catalog No. NC0015934 Supplier Lonza Walkersville Supplier No. LT27008
The ATP Standard is a specialized aqueous preparation of adenosine triphosphate (ATP) and is primarily intended for use in research to calibrate ATP assays based on the luciferase bioluminescence technique. Each vial contains 5 ml of 10 µM ATP.Content:1 x Bottle ATP Standard, 5 mL