Learn More
Revvity Health Sciences Inc ATP, [γ-32P]- 10Ci/mmol 2mCi/ml , 250 µCi
Supplier:  Revvity Health Sciences Inc BLU002250UC
Encompass
ATP, [γ-32P]- 10Ci/mmol 2mCi/ml , 250 µCi
Catalog No. 50-280-9323