Learn More
Medchemexpress LLC ATP synthase inhibitor 2 | 2814540-76-4 | 99.9% | 382.48 g/mol | C21H22N2O3S | 10 MG
SDP
Catalog No. 5000286756
Encompass_Preferred
ATP synthase inhibitor 2 is a research small-molecule inhibitor that targets the ATP synthase of Pseudomonas aeruginosa. It inhibits bacterial ATP synthesis (IC50 = 10 μg/mL) and can fully inhibit ATP synthesis at 128 μg/mL; however, it shows limited whole-cell antibiotic activity at the concentrations tested. The compound is provided as a solid for laboratory research with storage recommendations for powder and solutions.
Catalog No. 50-002-86756 Supplier Medchemexpress LLC Supplier No. HY15098310MG
ATP synthase inhibitor 2 is a research small-molecule inhibitor that targets the ATP synthase of Pseudomonas aeruginosa. It inhibits bacterial ATP synthesis (IC50 = 10 μg/mL) and can fully inhibit ATP synthesis at 128 μg/mL; however, it shows limited whole-cell antibiotic activity at the concentrations tested. The compound is provided as a solid for laboratory research with storage recommendations for powder and solutions.