Learn More
Medchemexpress LLC ATP synthase inhibitor 2 | 2814540-76-4 | C21H22N2O3S | 5 MG
SDP
Catalog No. 5000375896
Encompass_Preferred
This product is an ATP synthase inhibitor, specifically targeting *Pseudomonas aeruginosa*. It demonstrates complete inhibition of ATP synthesis activity in this bacterium at a concentration of 128 μg/mL.
  • Inhibits ATP synthase.
  • Specifically targets *Pseudomonas aeruginosa*.
  • Achieves complete inhibition of ATP synthesis at 128 μg/mL.
  • Available in solid form, white to off-white in color.
  • Has high purity.
Catalog No. 50-003-75896 Supplier Medchemexpress LLC Supplier No. HY1509835MG
This product is an ATP synthase inhibitor, specifically targeting *Pseudomonas aeruginosa*. It demonstrates complete inhibition of ATP synthesis activity in this bacterium at a concentration of 128 μg/mL.
  • Inhibits ATP synthase.
  • Specifically targets *Pseudomonas aeruginosa*.
  • Achieves complete inhibition of ATP synthesis at 128 μg/mL.
  • Available in solid form, white to off-white in color.
  • Has high purity.