Learn More
Gold Biotechnology Inc Adenosine-5'-Triphosphate (ATP), Disodium trihydrate, Ultra Pure 25 g
SDP
Catalog No. NC2941167
Encompass_Preferred
Adenosine-5'-triphosphate (ATP) is an important molecule in vivo for energy storage and metabolism and is often called the molecular unit of currency" of intracellular energy transfer. ATP is a coenzyme in multiple reactions as well as a substrate for multiple kinases where it helps to propagate a signal either inside or outside of the cell.
Catalog No. NC2941167 Supplier Gold Biotechnology Inc Supplier No. A08125
Adenosine-5'-triphosphate (ATP) is an important molecule in vivo for energy storage and metabolism and is often called the molecular unit of currency" of intracellular energy transfer. ATP is a coenzyme in multiple reactions as well as a substrate for multiple kinases where it helps to propagate a signal either inside or outside of the cell.