Learn More
ABclonal Technology ATP10A Rabbit pAb
SDP
Supplier:  ABclonal Technology A158731000UL
Encompass_Preferred
The protein encoded by this gene belongs to the family of P-type cation transport ATPases, and to the subfamily of aminophospholipid-transporting ATPases. The aminophospholipid translocases transport phosphatidylserine and phosphatidylethanolamine from one side of a bilayer to another. This gene is maternally expressed. It maps within the most common interval of deletion responsible for Angelman syndrome, also known as happy puppet syndrome.
Catalog No. 50-001-05041