Learn More
Aviva Systems Biology Corp Atp10d Antibody - middle region (ARP49515_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49515_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Atp10d. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Rat. Alternate names: Atp10dApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-432