Learn More
Aviva Systems Biology Corp ATP10D Antibody - C-terminal region (ARP49516_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49516_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ATP10D. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ATP10D. Alternate names: ATPVD, KIAA1487ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-53589