Learn More
Aviva Systems Biology Corp ATP11B Antibody - N-terminal region (ARP47046_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47046_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ATP11B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ATP11B. Alternate names: ATPIF, ATPIR, DKFZP434J238, DKFZP434N1615, KIAA0956, MGC46576ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59458