Learn More
ABclonal Technology ATP12A Rabbit pAb
SDP
Supplier:  ABclonal Technology A143721000UL
Encompass_Preferred
The protein encoded by this gene belongs to the family of P-type cation transport ATPases. This gene encodes a catalytic subunit of the ouabain-sensitive H+/K+ -ATPase that catalyzes the hydrolysis of ATP coupled with the exchange of H(+) and K(+) ions across the plasma membrane. It is also responsible for potassium absorption in various tissues. Two transcript variants encoding different isoforms have been found for this gene.
Catalog No. 50-001-07052