Learn More
Aviva Systems Biology Corp ATP1B1 Antibody - N-terminal region (ARP44410_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44410_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ATP1B1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ATP1B1. Alternate names: ATP1B, MGC1798ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-58801