Learn More
Aviva Systems Biology Corp ATP1B1 Antibody - middle region (ARP44411_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44411_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ATP1B1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human ATP1B1. Alternate names: ATP1B, MGC1798ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-014-53419