Learn More
Aviva Systems Biology Corp ATP1B4 Antibody - middle region (ARP46848_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46848_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ATP1B4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ATP1B4. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54028