Learn More
Aviva Systems Biology Corp ATP2A2 Antibody - C-terminal region (ARP45159_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45159_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ATP2A2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ATP2A2. Alternate names: ATP2B, DAR, DD, DKFZp686P0211, FLJ20293, FLJ38063, MGC45367, SERCA2ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58975