Learn More
Aviva Systems Biology Corp ATP2A3 Antibody - middle region (ARP46580_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46580_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ATP2A3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ATP2A3. Alternate names: SERCA3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rabbit, Rat
Catalog No. 50-014-59353