Learn More
Aviva Systems Biology Corp ATP5F1B Antibody - N-terminal region (ARP48185_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48185_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ATP5B. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human ATP5B. Alternate names: ATPMB, ATPSB, MGC5231ApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-59747