Learn More
Aviva Systems Biology Corp Atp5d Antibody - C-terminal region (ARP56323_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56323_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Atp5d. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 0610008F14Rik, 1500000I11Rik, AA960090, AI876556, AU020773, C85518ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-286-140