Learn More
ABclonal Technology ATP5G1 Rabbit mAb
SDP
Supplier:  ABclonal Technology A2096120UL
Encompass_Preferred
This gene encodes a subunit of mitochondrial ATP synthase Mitochondrial ATP synthase catalyzes ATP synthesis utilizing an electrochemical gradient of protons across the inner membrane during oxidative phosphorylation ATP synthase is composed of two linked multi-subunit complexes the soluble catalytic core F1 and the membrane-spanning component Fo comprising the proton channel The catalytic portion of mitochondrial ATP synthase consists of 5 different subunits (alpha beta gamma delta and epsilon) assembled with a stoichiometry of 3 alpha 3 beta and a single representative of the other 3 The proton channel seems to have nine subunits (a b c d e f g F6 and 8) This gene is one of three genes that encode subunit c of the proton channel Each of the three genes have distinct mitochondrial import sequences but encode the identical mature protein Alternatively spliced transcript variants encoding the same protein have been identified
Catalog No. 50-000-07953