Learn More
Aviva Systems Biology Corp ATP5MC2 Antibody - N-terminal region (ARP40228_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40228_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ATP5G2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ATP5G2. Alternate names: ATP5AApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-53179