Learn More
Aviva Systems Biology Corp ATP5PD Antibody - C-terminal region (ARP52160_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52160_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ATP5H. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human ATP5H. Alternate names: ATP5JD, ATPQApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-285-653