Learn More
ABclonal Technology ATP6 Rabbit mAb
SDP
Catalog No. 5000004887
Encompass_Preferred
Predicted to contribute to proton-transporting ATP synthase activity rotational mechanism Predicted to be involved in mitochondrial ATP synthesis coupled proton transport Located in mitochondrion Is expressed in several structures including alimentary system brain heart integumental system and sensory organ Human ortholog(s) of this gene implicated in Leber hereditary optic neuropathy NARP syndrome Parkinson s disease multiple sclerosis and systemic lupus erythematosus Orthologous to human MT-ATP6 (mitochondrially encoded ATP synthase membrane subunit 6)
Catalog No. 50-000-04887 Supplier ABclonal Technology Supplier No. A23150100UL
Predicted to contribute to proton-transporting ATP synthase activity rotational mechanism Predicted to be involved in mitochondrial ATP synthesis coupled proton transport Located in mitochondrion Is expressed in several structures including alimentary system brain heart integumental system and sensory organ Human ortholog(s) of this gene implicated in Leber hereditary optic neuropathy NARP syndrome Parkinson s disease multiple sclerosis and systemic lupus erythematosus Orthologous to human MT-ATP6 (mitochondrially encoded ATP synthase membrane subunit 6)