Learn More
ABclonal Technology ATP6AP1 Rabbit mAb
SDP
Catalog No. 5000007176
Encompass_Preferred
This gene encodes a component of a multisubunit enzyme that mediates acidification of eukaryotic intracellular organelles Vacuolar ATPase (V-ATPase) is comprised of a cytosolic V1 (site of the ATP catalytic site) and a transmembrane V0 domain V-ATPase dependent organelle acidification is necessary for such intracellular processes as protein sorting zymogen activation and receptor-mediated endocytosis The encoded protein of this gene may assist in the V-ATPase-mediated acidification of neuroendocrine secretory granules This protein may also play a role in early development
Catalog No. 50-000-07176 Supplier ABclonal Technology Supplier No. A2246820UL
This gene encodes a component of a multisubunit enzyme that mediates acidification of eukaryotic intracellular organelles Vacuolar ATPase (V-ATPase) is comprised of a cytosolic V1 (site of the ATP catalytic site) and a transmembrane V0 domain V-ATPase dependent organelle acidification is necessary for such intracellular processes as protein sorting zymogen activation and receptor-mediated endocytosis The encoded protein of this gene may assist in the V-ATPase-mediated acidification of neuroendocrine secretory granules This protein may also play a role in early development