Learn More
Aviva Systems Biology Corp ATP6V0A1 Antibody - N-terminal region (ARP46581_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46581_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ATP6V0A1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ATP6V0A1. Alternate names: ATP6N1, ATP6N1A, DKFZp781J1951, Stv1, VPP1, Vph1, a1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59354