Learn More
Aviva Systems Biology Corp ATP6V0A2 Antibody - N-terminal region (ARP42365_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42365_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ATP6V0A2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ATP6V0A2. Alternate names: ATP6N1D, ATP6a2, J6B7, Stv1, TJ6, TJ6M, TJ6s, Vph1, a2, A2, RTF, WSS, ARCL, STV1, TJ6S, VPH1, ARCL2A, ATP6A2ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat, Yeast, Zebrafish
Catalog No. 50-014-58201