Learn More
Aviva Systems Biology Corp ATP6V0C Antibody - middle region (ARP45161_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45161_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ATP6V0C. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ATP6V0C. Alternate names: ATP6C, ATP6L, ATPL, VATL, Vma3, VPPCApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rabbit, Rat, Sheep
Catalog No. 50-014-54027